missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Decahydrate, puriss., ≥99%, Honeywell™ Fluka™

Product Code. 15601360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15601360 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15601360 Supplier Honeywell-Fluka Supplier No. 135712.5KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, European Pharmacopoeia, USP,≥99.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 32.4°C
CAS 7727-73-3
Quantity 2.5 kg
Molecular Formula H20Na2O14S
Linear Formula Na2SO4 · 10H2O
MDL Number MFCD00149189
UN Number NONH for all modes of transport
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfate;decahydrate
Molecular Weight (g/mol) 322.186
PubChem CID 62649
ChEBI CHEBI:32586
Formula Weight 322.2g/mol
Percent Purity ≥99.0% (calc. to the dried substance)
Grade puriss.
Packaging Poly Bottle
Density 1.46g/mL at 25°C
Chemical Name or Material Sodium Sulfate Decahydrate, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.