missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate decahydrate, 99%

Product Code. 11467667
Change view
Click to view available options
Quantity:
1000 g
25,000 g
5000 g
Unit Size:
1000g
25000g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467667 5000 g 5000g
11457667 1000 g 1000g
11477667 25,000 g 25000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467667 Supplier Thermo Scientific Alfa Aesar Supplier No. A15702.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7727-73-3 | H20Na2O14S | 322.186 g/mol

Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate decahydrate is used as a laboratory reagent to precipitate protein. It is largely used as filler in powdered home laundry detergents. It facilitates in leveling, reducing negative charges on fibers, thereby allows dye to penetrate evenly. It avoids the corrosion of stainless steel vessels used in dyeing. It is also used as a laxative and effectively replaces the drug paracetamol in the body.

Solubility
Soluble in water and glycerin. Insoluble in ethanol and methanol.

Notes
Stable. Incompatible with strong acids, aluminum and magnesium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-73-3
Molecular Formula H20Na2O14S
Molecular Weight (g/mol) 322.186
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point ∼32°C
pH 5.2 to 8.0
Quantity 5000 g
Merck Index 14,8680
Solubility Information Soluble in water and glycerin. Insoluble in ethanol and methanol.
Formula Weight 322.19 (142.04 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 1.46
Chemical Name or Material Sodium sulfate decahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.