missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T. Baker™
414.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
45.5 kg
Packaging:
Glass Bottle
Lined Fiber Drum
Wide Mouth Amber Glass Bottle
Specifications
pH | 5.2 to 9.2 |
Physical Form | Granules |
Quantity | 2.5 kg |
IUPAC Name | disodium sulfate |
Formula Weight | 142.04 |
Grade | Meets ACS reagent chemical requirements |
Loss on Ignition | max. 0.5% |
Packaging | Glass Bottle |
Specific Gravity | 1L = 2.68kg |
Chemical Name or Material | Sodium Sulfate, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'