missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, anhydrous, 99%

Product Code. 11413970
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413970 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413970 Supplier Thermo Scientific Alfa Aesar Supplier No. A19890.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.

Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 884°C
CAS 7757-82-6
Quantity 500 g
Molecular Formula Na2O4S
Linear Formula Na2SO4
MDL Number MFCD00003504
Merck Index 14,8680
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous
Solubility Information Soluble aq. soln.,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium sulfate
Molecular Weight (g/mol) 142.04
PubChem CID 24436
ChEBI CHEBI:32149
Formula Weight 142.04
Percent Purity 99%
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Density 2.68
Chemical Name or Material Sodium sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.