missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
23.09 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1000 g
5000 g
Description
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
Specifications
Melting Point | 884°C |
Quantity | 1000 g |
Linear Formula | Na2SO4 |
Merck Index | 14,8680 |
Solubility Information | Soluble in water,glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents. |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.484 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'