missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15698140 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
10 kg
Packaging:
Fiber drum
Poly bottle
Unit Size:
10kg
2.5kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15698140

Brand: Honeywell Research Chemicals S9627500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Melting Point 884°C
Color Colorless
Physical Form Solid
Quantity 500 g
Linear Formula Na2SO4
UN Number NONH for all modes of transport
Formula Weight 142.04g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Density 2.68g/mL at 25°C
Chemical Name or Material Sodium sulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.