missing translation for 'onlineSavingsMsg'
Learn More

Sodium sticks, coated in film of protective hydrocarbon oil, 99%

Product Code. 11416800
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416800 100 g 100g
11426800 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416800 Supplier Thermo Scientific Alfa Aesar Supplier No. L13285.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-23-5 | Na | 22.99 g/mol

Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]

Specifications

Melting Point 98°C
Boiling Point 881°C
Quantity 100 g
Flash Point >110°C (230°F)
UN Number UN1428
Merck Index 14,8570
Percent Purity 99%
Chemical Name or Material Sodium sticks, in mineral oil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.