missing translation for 'onlineSavingsMsg'
Learn More

Sodium stearate

Product Code. 11384597
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 11384597

Brand: Thermo Scientific Alfa Aesar 039648.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-16-2 | C18H35NaO2 | 306.466 g/mol

Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-16-2
Molecular Formula C18H35NaO2
Molecular Weight (g/mol) 306.466
MDL Number MFCD00036404
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
Synonym sodium stearate, sodium octadecanoate, octadecanoic acid, sodium salt, stearates, stearic acid, sodium salt, prodhygine, flexichem b, stearic acid sodium salt, bonderlube 235, unii-qu7e2xa9tg
PubChem CID 2724691
IUPAC Name sodium;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].[Na+]

Specifications

Odor Weak
Quantity 5 kg
Merck Index 14,8678
Solubility Information Slowly soluble in cold water.
Formula Weight 306.46
Physical Form Powder
Chemical Name or Material Sodium stearate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.