missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard for Atomic Absorption (AAS) 1000 μg/mL (1000 ppm) in 0.5 M Nitric Acid (HNO3), Reagecon
BIOPHARMA

Product Code. 11489129 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489129 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11489129 Supplier Reagecon Supplier No. AANAH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured gravimetrically from very pure metal or salt (at least 99.9%) and tested by an ISO17025 Accredited Test Method (INAB Ref: 264T).

  • Atomic Absorption Reference Standard, Atomic Absorption Reference Material
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of −196 to +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), high quality acids and pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.

Specifications

Description Matrix: 0.5 M Nitric Acid (HNO3)
Packaging HDPE Triple-leached
Analyte or Component Names Sodium
Physical Form Liquid
Concentration 1000 μg/mL (1000 ppm)
Element Na
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • SODIUM AAS STANDARD SOLUTION
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, sodiumnitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.