missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate, Technical, Solution, d=1.5, Fisher Chemical™

Product Code. 10292052 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10292052 25 L Plastic container 25 L
10060470 2.5 L HDPE plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10292052 Supplier Fisher Chemical Supplier No. S/6340/25
  • ONLINE EXCLUSIVE PRICE
    £406.76
    £363.00 / 25 L
    Save £43.76
    10% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Silicate, Technical, Solution, d=1.5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Boiling Point 100.6°C
Melting Point -3°C
Color Colorless
pH 11.2
Physical Form Liquid
Quantity 25 L
Packaging Plastic container
Density Vapor Density: 0.7
compliance-icons compliance-icons
Product Identifier
  • Sodium silicate, solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.