missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate Solution, Reagent Grade, Solstice

Product Code. 15674090 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
3 L
Packaging:
Poly bottle
Unit Size:
1 L
25mL
3 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15674090 3 L Poly bottle 3 L
15618370 25 mL Poly bottle 25mL
15664090 1 L Poly bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15674090 Supplier Honeywell Research Chemicals Supplier No. 3384433L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Quantity 3 L
Linear Formula Na2O(SiO2)x · xH2O
UN Number UN3266
Packaging Poly bottle
Density 1.39g/mL at 25°C
Chemical Name or Material Sodium silicate solution
compliance-icons compliance-icons
Product Identifier
  • Sodium silicate solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium silicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.