missing translation for 'onlineSavingsMsg'
Learn More

Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™

Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds

Brand:  Thermo Scientific Alfa Aesar 044893.14

Code : NEW

Additional Details : Weight : 0.02500kg Transport : UN number : 2813 Chem class : 4.3 Pack group : II

 View more versions of this product

Product Code. 15466625

  • £204.00 / 25g
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sodium-silica gel
Black
25 g
Reacts with water.
0.7 g/cm3
35-40% alkali metal in silica gel, Stage I
Powder
MFCD01941718
Moisture sensitive
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers