missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite Pentahydrate, TRC

Product Code. 30180257
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180257 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180257 Supplier Toronto Research Chemicals Supplier No. TRCS67090050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 26970-82-1
Quantity 50 g
Molecular Formula H10Na2O8Se
Synonym Selenious Acid Disodium Salt Pentahydrate, Selenious Acid Disodium Salt Hydrate
InChI Formula InChI=1S/2Na.H2O3Se.5H2O/c;;1-4(2)3;;;;;/h;;(H2,1,2,3);5*1H2/q2*+1;;;;;;/p-2
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Se](=O)[O-]
IUPAC Name disodium;selenite;pentahydrate
Molecular Weight (g/mol) 263.01
Formula Weight 263.9336
Chemical Name or Material Sodium Selenite Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.