missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite Pentahydrate, TRC

Product Code. 30180139
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180139 100 g 100g
30180076 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180139 Supplier Toronto Research Chemicals Supplier No. TRCS670900100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26970-82-1
Molecular Formula H10Na2O8Se
Molecular Weight (g/mol) 263.01
Synonym Selenious Acid Disodium Salt Pentahydrate, Selenious Acid Disodium Salt Hydrate
IUPAC Name disodium;selenite;pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Se](=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2Na.H2O3Se.5H2O/c;;1-4(2)3;;;;;/h;;(H2,1,2,3);5*1H2/q2*+1;;;;;;/p-2
Formula Weight 263.9336
Chemical Name or Material Sodium Selenite Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.