missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis)

Product Code. 11376449
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376449 500 g 500g
11366449 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376449 Supplier Thermo Scientific Alfa Aesar Supplier No. 012585.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In glass manufacturing | CAS: 10102-18-8 | Na2O3Se | 172.948 g/mol

In glass manufacturingSodium selenite is used in the manufacture of colorless glass. It acts as an ingredient in some food supplements. It is an animal feed used in pet foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.948
MDL Number MFCD00003489
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
PubChem CID 24934
ChEBI CHEBI:48843
IUPAC Name disodium;selenite
SMILES [O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Melting Point >350°C
Color White
pH 9
Physical Form Powder
Quantity 500 g
Linear Formula Na2SeO3
UN Number UN2630
Merck Index 14,8673
Solubility Information Freely soluble in water; Insoluble in alcoholSoluble in water; Insoluble in alcohol and ethanol.
Formula Weight 172.94
Percent Purity ≤99%
Odor Odorless
Sensitivity Hygroscopic
Assay Typically ≤99.75% (metals basis)
Chemical Name or Material Sodium selenite, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.