missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenite, 44-46% Se, anhydrous

Product Code. p-3735337 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10318340

Brand: Thermo Scientific 200730250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-18-8 | Na2O3Se | 172.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.94
MDL Number MFCD00003489
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
PubChem CID 24934
ChEBI CHEBI:48843
IUPAC Name disodium;selenite
SMILES [O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 350°C
Color White to Beige
Physical Form Powder
Quantity 25 g
Linear Formula Na2SeO3
Merck Index 15, 8802
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 172.94
Percent Purity 44 to 46% Se
Loss on Drying 0.5% max.
Packaging Glass bottle
Chemical Name or Material Sodium selenite, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium selenite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.