missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenate, MP Biomedicals

Product Code. 11422085 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
11422085 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 11422085

Brand: MP Biomedicals 0520364950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 13410-01-0
Quantity 50 g
Molecular Formula Na2O4Se
MDL Number MFCD00003490
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O
IUPAC Name disodium selenate
Molecular Weight (g/mol) 188.95
PubChem CID 25960
ChEBI CHEBI:77775
Formula Weight 188.94
Chemical Name or Material Sodium Selenate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.