missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenate Hydrate, TRC

Product Code. 30419535
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419535 250 mg 250mg
30419035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30419535 Supplier Toronto Research Chemicals Supplier No. TRCS224760250MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Selenate Hydrate
CAS 10102-23-5
Quantity 250 mg
Molecular Formula 10 H2 O . 2 Na . O4 Se
Synonym Sodium Selenate Hydrate, Sodium Selenate (Na2SeO4) Decahydrate
InChI Formula InChI=1S/2Na.H2O4Se.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se](=O)(=O)[O-]
IUPAC Name disodium;selenate;decahydrate
Molecular Weight (g/mol) 369.09
Formula Weight 369.981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.