missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate decahydrate, 99.9% (metals basis)

Product Code. p-7049404
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11316479

Brand: Thermo Scientific Alfa Aesar 014239.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-23-5 | H20Na2O14Se | 369.10 g/mol

Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-23-5
Molecular Formula H20Na2O14Se
Molecular Weight (g/mol) 369.10
MDL Number MFCD00149174
InChI Key TYVNFQRMDPFYPC-UHFFFAOYSA-L
Synonym sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis
PubChem CID 202320
IUPAC Name disodium;selenate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Melting Point 35°C (decomposition)
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
UN Number UN2630
Merck Index 14,8671
Solubility Information Soluble in water.
Formula Weight 369.09 (188.94 Anhydrous)
Percent Purity 99.9%
Density 1.6 g/mL
Assay (metals basis)
Chemical Name or Material Sodium selenate decahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.