missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate decahydrate, 99.9% (metals basis)

Product Code. 11326479
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326479 100 g 100g
11316479 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326479 Supplier Thermo Scientific Alfa Aesar Supplier No. 014239.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-23-5 | H20Na2O14Se | 369.10 g/mol

Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-23-5
Molecular Formula H20Na2O14Se
Molecular Weight (g/mol) 369.10
MDL Number MFCD00149174
InChI Key TYVNFQRMDPFYPC-UHFFFAOYSA-L
Synonym sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis
PubChem CID 202320
IUPAC Name disodium;selenate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Melting Point 35°C (decomposition)
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
UN Number UN2630
Merck Index 14,8671
Solubility Information Soluble in water.
Formula Weight 369.09 (188.94 Anhydrous)
Percent Purity 99.9%
Density 1.600 g/mL
Assay (metals basis)
Chemical Name or Material Sodium selenate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.