missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Description
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 32° (transition) |
Color | Purple |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.8+% (metals basis) |
Solubility Information | Soluble in water. |
Formula Weight | 188.94 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White powder |
Show More |
Safety and Handling
Product Identifier
- Sodium selenate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H300-Fatal if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P330-Rinse mouth.
- MIXTURE LIST-Contains : sodium selenate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'