missing translation for 'onlineSavingsMsg'
Learn More

Sodium selenate, 98%

Product Code. 10095193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095193 100 g Glass bottle 100g
10177083 25 g Glass bottle 25g
10278653 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095193 Supplier Thermo Scientific Acros Supplier No. 369961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
IUPAC Name disodium;selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 97.5% min
Merck Index 15, 8800
Solubility Information Solubility in water: 19g/L (20°C)
Formula Weight 188.94
Percent Purity 98%
Identification confirmed
Packaging Glass bottle
Chemical Name or Material Sodium selenate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium selenate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.