missing translation for 'onlineSavingsMsg'
Learn More

Sodium Salicylate, MedChemExpress

Product Code. 16448378
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448378 10 g 10g
16454267 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448378 Supplier Medchem Express Supplier No. HYB0167A
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium Salicylate (Salicylic acid sodium salt) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. Sodium Salicylate is also a S6K inhibitor.Sodium Salicylate is a NF-κB inhibitor that decreases inflammatory gene expression and improves repair in aged muscle.

Sodium Salicylate (Salicylic acid sodium salt) inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation. Sodium Salicylate is also a S6K inhibitor.Sodium Salicylate is a NF-κB inhibitor that decreases inflammatory gene expression and improves repair in aged muscle.

Specifications

Chemical Name or Material Sodium Salicylate
CAS 54-21-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H5NaO3
Quantity 10 g
Synonym Salicylic acid sodium salt2-Hydroxybenzoic acid sodium salt
Solubility Information H2O : ≥ 100 mg/mL (624.61 mM) ∣DMSO : 100 mg/mL (624.61 mM; Need ultrasonic)
SMILES O=C(O[Na])C1=CC=CC=C1O
Molecular Weight (g/mol) 160.1
Formula Weight 160.1
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.