missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate tetrabasic decahydrate, Puriss. p.a., ≥99%, Honeywell Fluka™

Product Code. p-7071966 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
6 x 2.5 kg
Unit Size:
2.5kg
500g
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15653020 500 g 500g
15643020 2.5 kg 2.5kg
15673020 6 x 2.5 kg Pack of 6
15663020 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15653020 Supplier Honeywell-Fluka Supplier No. 30411500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na4P2O7 · 10H2O
MDL Number MFCD00149200
UN Number NONH for all modes of transport
Formula Weight 446.06g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Chemical Name or Material Sodium Pyrophosphate Tetrabasic Decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.