missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Dibasic, TRC

Product Code. 30423197
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423197 Supplier Toronto Research Chemicals Supplier No. TRCS0839331G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Pyrophosphate Dibasic
CAS 7758-16-9
Molecular Formula H2Na2O7P2
Quantity 1 g
Synonym Disodium Diphosphate, Pyrophosphoric Acid Disodium Salt, Diphosphoric Acid Disodium Salt, Disodium Pyrophosphate, Disodium Acid Pyrophosphate
InChI Formula InChI=1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2
SMILES [Na+].[Na+].OP(=O)([O-])OP(=O)(O)[O-]
IUPAC Name disodium;[hydroxy(oxido)phosphoryl] hydrogen phosphate
Molecular Weight (g/mol) 221.94
Formula Weight 221.9071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.