Learn More
Sodium pyrophosphate decahydrate, 99+%, for analysis, ACROS Organics™
Brand: Acros Organics 205970250
Code : 29
Additional Details : CAS Number : 13472-36-1 Weight : 0.07500kg
Quantity | 25g |
---|
Description
Biological bufferChemical Identifiers
13472-36-1 | |
446.05 | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
3084150 | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
Na4O7P2·10H2O | |
MFCD00149200 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
tetrasodium;phosphonato phosphate;decahydrate |
Specifications
Sodium pyrophosphate decahydrate | |
13472-36-1 | |
99+% | |
Na4P2O7·10H2O | |
10,9063 | |
Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia,insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
446.05 | |
446.05 | |
99+% | |
10ppm max | |
1.82 | |
5ppm max. | |
50ppm max. | |
93.8°C | |
80.0°C |
p.a. | |
ortho-phosphate (as PO4): < 0.1% | |
Na4O7P2·10H2O | |
MFCD00149200 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
tetrasodium;phosphonato phosphate;decahydrate | |
3084150 | |
Crystals | |
Analysis | |
10ppm max. | |
5ppm max. | |
Glass Bottle | |
0.001% max. (as N) | |
White | |
25g |
Safety and Handling
- Sodium pyrophosphate decahydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium pyrophosphatedecahydrate, Sodiumpyrophosphate