missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate decahydrate, 99-103%, ACS reagent

Product Code. 12387073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12387073 500 g 500g
12327203 100 g 100g
12317203 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 12387073 Supplier Thermo Scientific Acros Supplier No. 448065000
  • ONLINE EXCLUSIVE PRICE
    £45.15
    £43.80 / 500g
    Save £1.35
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

CAS: 13472-36-1 | Na4O7P2·10H2O | 446.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-36-1
Molecular Formula Na4O7P2·10H2O
Molecular Weight (g/mol) 446.05
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Boiling Point 93.8°C
Melting Point 80.0°C
Quantity 500 g
Assay Percent Range 99-103%
Linear Formula Na4P2O7·10H2O
Solubility Information Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia, insoluble in alcohol
Formula Weight 446.05
Percent Purity 99 to 103%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium pyrophosphate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.