missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyrophosphate decahydrate, 97+%

Product Code. 11429210
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429210 250 g 250g
11409210 1000 g 1000g
11419210 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429210 Supplier Thermo Scientific Alfa Aesar Supplier No. A16195.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13472-36-1 | H20Na4O17P2 | 446.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.

Solubility
Soluble in water. Insoluble in alcohol and ammonia.

Notes
Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
MDL Number MFCD00149200
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 79.5°C
pH 9.5 to 10.5
Quantity 250 g
Linear Formula Na4P2O7·10H2O
Merck Index 14,9240
Solubility Information Soluble in water. Insoluble in alcohol and ammonia.
Formula Weight 446.06 (265.90 Anhydrous)
Percent Purity ≥97%
Odor Odorless
Density 1.82
Chemical Name or Material Sodium pyrophosphate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.