missing translation for 'onlineSavingsMsg'
Learn More

Sodium propionate, 99%

Product Code. 11478027
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478027 2500 g 2500g
11468027 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478027 Supplier Thermo Scientific Alfa Aesar Supplier No. A17440.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Vea la política de devoluciones

CAS: 137-40-6 | C3H5NaO2 | 96.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium propionate is used as a food preservative, as a mold inhibitor in bakery products and in insect cell culture. It is also used in pharmaceuticals, cosmetics, agriculture and animal feed as preservative. It acts as a food additive. Further, it inhibits the growth of lactic acid bacteria.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.061
MDL Number MFCD00002759
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
PubChem CID 2723816
IUPAC Name sodium;propanoate
SMILES CCC(=O)[O-].[Na+]

Especificaciones

Melting Point 285°C to 291°C
Assay Percent Range 99%
Linear Formula CH3CH2CO2Na
Quantity 2500 g
Sensitivity Hygroscopic
Merck Index 14,8669
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 96.06
Percent Purity 99%
Chemical Name or Material Sodium propionate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.