Learn More
DiscoverX Sodium propionate
Product Code. 17332529
Encompass
Click to view available options
:
10 mg
Product Code. 17332529
Brand: DiscoverX 921205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sodium propionate is an agonist for FFAR2 and FFAR3. Provided as 10mg of dry powder.
Sodium propionate is an agonist for FFAR2 and FFAR3. Also used as food additive preservative against mold and bacteria. It is the salt form of propionic acid, an organic acid produced during sugar degradation, but is also a commonly synthesized molecule. Provided as 10mg of dry powder.
Specifications
Quantity 10 mg