missing translation for 'onlineSavingsMsg'
Learn More

Sodium Prasterone Sulfate, TRC

Product Code. 30424219
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424219 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424219 Supplier Toronto Research Chemicals Supplier No. TRCS673250100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydroepiandrosterone 3-sulfate sodium salt (DHEAS) (as free sulfate)
CAS 1099-87-2
Molecular Formula C19 H27 O5 S . Na
Quantity 100 mg
Synonym Androst-5-en-17-one','3-(sulfooxy)-','sodium salt (1:1)','(3β)-, Androst-5-en-17-one','3-(sulfooxy)-','sodium salt','(3β)- (9CI), Androst-5-en-17-one','3β-hydroxy-','hydrogen sulfate','sodium salt (6CI,7CI,8CI), DHA-S, DHEA sulfate sodium salt, Dehydroepiandrosterone sulfate sodium salt, Dehydroisoandrosterone sulfate sodium salt, Mylis, NSC 72822, Prasterone sodium sulfate, Sodium dehydroepiandrosterone sulfate, Sodium dehydroepiandrosterone-3-sulfate, Sodium prasterone sulfate, Teloin
InChI Formula InChI=1S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14-,15-,16-,18-,19-;/m0./s1
SMILES [Na+].C[C@]12CC[C@H]3[C@@H](CC=C4C[C@H](CC[C@]34C)OS(=O)(=O)[O-])[C@@H]1CCC2=O
IUPAC Name sodium;[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-oxo-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-yl] sulfate
Molecular Weight (g/mol) 390.47
Formula Weight 390.1477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.