missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium Tartrate Tetrahydrate, ≥99%, MP Biomedicals™

Product Code. 11476602 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476602 1 kg 1kg
11486602 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476602 Supplier MP Biomedicals Supplier No. 0215015601
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
MDL Number MFCD00150989
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range ≥99%
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Molecular Weight (g/mol) 282.218
Formula Weight 282.2
Percent Purity ≥99%
Chemical Name or Material Sodium Potassium Tartrate Tetrahydrate
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.