missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium ATPase Beta 1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18355676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355676 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355676 Supplier Novus Biologicals Supplier No. NBP300261DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sodium Potassium ATPase Beta 1 Polyclonal specifically detects Sodium Potassium ATPase Beta 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium Potassium ATPase Beta 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Sodium Potassium ATPase Beta 1 (P05026-1; Glu63-Ser303).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 481
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.