missing translation for 'onlineSavingsMsg'
Learn More

Sodium Potassium ATPase Alpha 3 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18381472 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381472 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381472 Supplier Novus Biologicals Supplier No. NBP299238AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sodium Potassium ATPase Alpha 3 Polyclonal specifically detects Sodium Potassium ATPase Alpha 3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium Potassium ATPase Alpha 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of mouse Sodium Potassium ATPase Alpha 3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 478
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.