missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 2 Antibody (JE34-00), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32061
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 2 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Sodium Potassium ATPase Alpha 2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 | |
| alpha-A(+) catalytic polypeptide, ATPase, Na+/K+ transporting, alpha 2 (+) polypeptide, ATPase, Na+/K+ transporting, alpha 2 polypeptide, EC 3.6.3, EC 3.6.3.9, KIAA0778, MGC59864, MHP2, migraine, hemiplegic 2, Na(+)/K(+) ATPase alpha-2 subunit, Na+/K+ ATPase, alpha-B polypeptide, Sodium pump subunit alpha-2, sodium/potassium-transporting ATPase alpha-2 chain, sodium/potassium-transporting ATPase subunit alpha-2, sodium-potassium ATPase | |
| Synthetic peptide within Human Sodium Potassium ATPase Alpha 2 aa 50-99/ 1,020. (Uniprot: P50993) | |
| 100 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 477 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE34-00 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction