missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 1 Antibody (464.6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 97 publications
£207.00 - £500.00
Specifications
| Antigen | Sodium Potassium ATPase Alpha 1 |
|---|---|
| Clone | 464.6 |
| Dilution | Western Blot 1:1000-1:10000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation reported in scientific literature (PMID 27748972), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 26941236) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18775013
|
Novus Biologicals
NB300-146SS |
0.025 mL |
£207.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789173
|
Novus Biologicals
NB300-146 |
0.05 mL |
£500.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium Potassium ATPase Alpha 1 Monoclonal specifically detects Sodium Potassium ATPase Alpha 1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Drosophila, Guinea Pig, Primate, Rabbit, Sheep, Xenopus, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Sodium Potassium ATPase Alpha 1 | |
| Western Blot 1:1000-1:10000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation reported in scientific literature (PMID 27748972), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 26941236) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Drosophila, Guinea Pig, Primate, Rabbit, Xenopus species, Yeast | |
| P05023 | |
| 476 | |
| Purified Sodium Potassium ATPase Alpha 1 from rabbit renal outer medulla. [UniProt# Q9N0Z6] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 112 kDa |
| 464.6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| Ascites with 0.1% Sodium Azide | |
| ATPase, Na+/K+ transporting, alpha 1 polypeptide, EC 3.6.3, EC 3.6.3.9, K-ATPase alpha-1 subunit, K-ATPase catalytic subunit alpha-A protein, MGC3285, MGC51750, Na, Na(+)/K(+) ATPase alpha-1 subunit, Na, K-ATPase, alpha-A catalytic polypeptide, Na+, K+ ATPase alpha subunit, Na+/K+ ATPase 1, sodium pump 1, Sodium pump subunit alpha-1, sodium/potassium-transporting ATPase subunit alpha-1, sodium-potassium-ATPase, alpha 1 polypeptide | |
| ATP1A1 | |
| IgG1 κ | |
| Unpurified | |
| This is specific for Na,K-ATPase alpha 1 subunit. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title