missing translation for 'onlineSavingsMsg'
Learn More

Sodium polyanetholesulfonate

Product Code. 10413171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10413171 100 g 100g
10667391 25 g 25g
11914721 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10413171 Supplier Thermo Scientific Acros Supplier No. 419631000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55963-78-5 | (C10H11NaO4S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55963-78-5
Molecular Formula (C10H11NaO4S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00148427
InChI Key JKJBFNAERWARKW-CZEFNJPISA-L
Synonym sodium anethole sulfite, polyanetholesulfonate, polyanethol sulfonate, benzene, 1-methoxy-4-1-propenyl-, e-, homopolymer, sulfonated, sodium salt, sodium 1-methoxy-4-e-prop-1-enyl benzene sulfite
PubChem CID 6434512
SMILES CS(=O)(=O)O[Na].COC1=CC=C(C=C1)C(-*)C(C)-*

Specifications

Color Tan to White
pH 5.0 to 7.0 (1% aq. soln.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Merck Index 14,8666
Solubility Information Solubility in water: soluble.
IUPAC Name sodium;1-methoxy-4-[(E)-prop-1-enyl]benzene;sulfite
Physical Form Crystals or Powder
Chemical Name or Material Sodium polyanetholesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.