missing translation for 'onlineSavingsMsg'
Learn More

Sodium pivalate hydrate, 98%

Product Code. 10729334 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10729334

Brand: Thermo Scientific Acros 364710500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143174-36-1 | C5H9NaO2 | 124.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143174-36-1
Molecular Formula C5H9NaO2
Molecular Weight (g/mol) 124.12
MDL Number MFCD00150782
InChI Key SJRDNQOIQZOVQD-UHFFFAOYSA-M
Synonym sodium trimethylacetate hydrate, sodium pivalate hydrate, sodium hydrate pivalate, propanoic acid, 2,2-dimethyl-, sodium salt, hydrate, acmc-20ajn9, sodiumtrimethylacetatehydrate, c5h9o2.na.h2o, trimethylacetic acid sodium salt hydrate, sodium 2,2-dimethylpropanoate-water 1/1/1
PubChem CID 23666172
IUPAC Name sodium;2,2-dimethylpropanoate;hydrate
SMILES [Na+].CC(C)(C)C([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)3CCO2Na·xH2O
Quantity 50 g
Formula Weight 142.13
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Sodium pivalate hydrate
compliance-icons
Product Identifier
  • Sodium pivalate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.