missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, tribasic, pure, anhydrous

Product Code. 10648044 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Plastic Drum
Unit Size:
10kg
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10648044

Brand: Thermo Scientific Acros 389810100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7601-54-9 | Na3O4P | 163.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.94
MDL Number MFCD00003510
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
Synonym trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
PubChem CID 24243
ChEBI CHEBI:37583
IUPAC Name trisodium;phosphate
SMILES [O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 kg
Linear Formula Na3PO4
Merck Index 15, 8792
Solubility Information Solubility in water: 88g/L (20°C)
Formula Weight 163.94
Percent Purity ≥96.0%
Grade Pure
Packaging Fiber Drum
Chemical Name or Material Sodium phosphate, tribasic, Anhydrous, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Sodium phosphate, tribasic
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.