missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Tribasic Dodecahydrate, Honeywell Fluka™

Product Code. 15622180 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15622180

Brand: Honeywell-Fluka 222003500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.119
MDL Number MFCD00149198
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na3PO4 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 380.12g/mol
Percent Purity ≥98%
Grade ACS Reagent
Density 1.62g/mL at 25°C
Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.