missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, tribasic, anhydrous, tech.

Product Code. 11336778
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336778 5 kg 5kg
11326778 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336778 Supplier Thermo Scientific Alfa Aesar Supplier No. 013438.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7601-54-9 | Na3O4P | 163.939 g/mol

Sodium phosphate, tribasic is used in soaps, detergents, photographic developers, water softeners and boiler scale removers. It is used as a food additive, as cleaning agents, as a protein precipitant and an active ingredient in fluxes designed to deoxygenate nonferrous metals for casting. It is used to prepare chlorinated trisodium phosphate by reacting with sodium hypochlorite and used as a disinfectant and bleach. Further, it is used in dairy substitutes and to inhibit mold. In addition, it is employed in ceramic production. It is also used for the preservation of biologically active soil samples and fractionization of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.939
MDL Number MFCD00003510
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
Synonym trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
PubChem CID 24243
ChEBI CHEBI:37583
IUPAC Name trisodium;phosphate
SMILES [O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Solubility Information Soluble in water. Insoluble in ethanol and carbon disulfide.
Formula Weight 163.94
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Chemical Name or Material Sodium phosphate, tribasic, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.