missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Monohydrate, TRC

Product Code. 30137062
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137062 5 g 5g
30136724 25 g 25g
30003915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137062 Supplier Toronto Research Chemicals Supplier No. TRCS6670455G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula Na . H2 O . H2 O4 P
Molecular Weight (g/mol) 137.99
Synonym Sodium dihydrogen phosphate monohydrate
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(=O)(O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1
Formula Weight 137.9694
Percent Purity >95%
Chemical Name or Material Sodium dihydrogen phosphate monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.