missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Monobasic, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™
£357.00 - £910.00
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 137.99 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
IUPAC Name | sodium;dihydrogen phosphate;hydrate |
SMILES | O.[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15388491
|
Avantor J.T.Baker
3802-01 |
500 g | Wide Mouth Amber Glass Bottle |
£357.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15528174
|
Avantor J.T.Baker
3802-05 |
2.5 kg | Poly Bottle |
£910.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
10049-21-5 | |
137.99 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 | |
O.[Na+].OP(O)(O)=O |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
sodium;dihydrogen phosphate;hydrate |
Specifications
10049-21-5 | |
MFCD00149208 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
Multi-Compendial/USP | |
Sodium Phosphate, Monobasic, Monohydrate |
H5NaO5P | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O | |
137.99 | |
137.99 | |
1L = 2.04kg |