missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Monobasic, Monohydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£337.00 - £980.00
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 139.00 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
IUPAC Name | sodium;dihydrogen phosphate;hydrate |
SMILES | O.[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15539670
|
Avantor J.T.Baker
4011-01 |
500 g | Poly Bottle |
£337.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15558204
|
Avantor J.T.Baker
4011-05 |
2.5 kg | Poly Bottle |
£980.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Liquid Chromatography and Molecular Biology applications
Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 | |
O.[Na+].OP(O)(O)=O |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
sodium;dihydrogen phosphate;hydrate |
Specifications
10049-21-5 | |
H5NaO5P | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
1L = 2.04kg |
Crystals | |
MFCD00149208 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
Ultrapure Bioreagent | |
Sodium Phosphate, Monobasic, Monohydrate |