missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Phosphate Monobasic, Monohydrate

Product Code. 30018786
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018786

Brand: Melford S23120500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Phosphate Monobasic Monohydrate is a chemical compound that is commonly known as monosodium phosphate monohydrate or sodium dihydrogen phosphate monohydrate.

In the life science field, it is primarily used as a buffering agent and as a source of phosphate ions. Buffers help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for enzymatic reactions, cell culture, and other molecular biology techniques.

Sodium Phosphate Monobasic Monohydrate is commonly employed in laboratory settings for the preparation of buffer solutions with specific pH ranges and buffering capacities. It is particularly useful in applications where a lower pH range is desired, as it has a buffering capacity in the acidic pH range.

Moreover, the compound is used in various biological and biochemical assays, protein purification processes, and the formulation of diagnostic kits and pharmaceutical products. It serves as a source of phosphate ions, which are essential for numerous biological processes such as DNA and RNA synthesis, energy metabolism, and signal transduction.

Specifications

Chemical Name or Material Sodium Phosphate Monobasic, Monohydrate
CAS 10049-21-5
Quantity 500 g
Molecular Formula NaH2PO4-H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.