missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic dihydrate, 99+%, for analysis

Product Code. 10339150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339150 2.5 kg Plastic Bottle 2.5kg
10227070 1 kg Plastic Bottle 1kg
10338493 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10339150 Supplier Thermo Scientific Acros Supplier No. 271750025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13472-35-0 | H2NaO42H2O | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H2NaO42H2O
Molecular Weight (g/mol) 156.01
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
SMILES O.O.OP(=O)(O)[O-].[Na+]

Specifications

Melting Point 60.0°C
Color White
Physical Form Adhering Crystals
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula NaH2PO4·2H2O
Merck Index 15, 8790
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 156.01
Percent Purity ≥99%
Grade Analytical
Loss on Drying 24% max.
Packaging Plastic Bottle
Chemical Name or Material Sodium phosphate, monobasic dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.