missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™

Product Code. 10133153
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133153 500 g Glass Bottle 500g
10173103 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133153 Supplier Fisher BioReagents Supplier No. BP329500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For tissue culture and molecular biology applications

Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
SMILES [Na+].OP(O)([O-])=O

Specifications

Boiling Point 203.9°C
Melting Point 100°C
Color White
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99 %
Merck Index 15, 8790
IUPAC Name sodium;dihydrogen phosphate
Formula Weight 119.98
Percent Purity ≥99%
Identification Pass Test
Packaging Glass Bottle
Density 2.04g/cm³
Chemical Name or Material Sodium Phosphate Monobasic Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.