missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic, 99%, for analysis, anhydrous

Product Code. 11914291 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 11914291

Brand: Thermo Scientific Acros 389870100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
SMILES [Na+].OP(O)([O-])=O

Specifications

Color White
pH 4.1 to 4.7 (5% soln.)
Physical Form Crystalline Powder
Quantity 10 kg
Assay Percent Range 99%
Linear Formula NaH2PO4
Merck Index 15,879
Solubility Information Solubility in water: 850g/L (20°C).
IUPAC Name sodium dihydrogen phosphate
Formula Weight 119.98
Percent Purity 99%
Grade Analytical
Packaging Plastic drum
Chemical Name or Material Sodium phosphate, monobasic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.