missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 250 g |
Assay Percent Range | 99% |
Linear Formula | NaH2PO4 |
Merck Index | 15,879 |
Solubility Information | (1 M in water) Clear colorless |
Formula Weight | 119.98 |
Percent Purity | 99% |
Grade | Biochemical |
Packaging | Plastic bottle |
Chemical Name or Material | Sodium phosphate, monobasic |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'