missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dodecahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number 11327327
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10101-89-0
H24Na3O16P
380.119
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10101-89-0
H24Na3O16P
380.119
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Sodium phosphate dodecahydrate is used in photographic developing, clarifying sugar, removing boiler scale, softening water and manufacturing of paper. They are added to many foods as an emulsifier to prevent oil separation. It can be utilized as a leavening agent. Some instances of these foods include the batter coating on breaded fish or chicken, and commercially baked cakes. Adding this item to food increases the shelf life of the food, maintaining the texture and visual aspect of the food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼75°C (decomposition)
Color White
pH 12
Physical Form Crystalline
Quantity 500 g
Linear Formula Na3PO4·12H2O
UN Number UN3262
Merck Index 14,8662
Solubility Information Soluble in water; Insoluble in alcohol.
Formula Weight 380.12 (163.94 Anhydrous)
Percent Purity 98.0 to 102.0%
Grade ACS
Odor Odorless
Density 1.62 g/mL
Chemical Name or Material Sodium phosphate dodecahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium phosphate dodecahydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'